Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
-
Chromosome Location
Chr.7: 148698178 - 148801110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003592.2 NP_003583.2 10 - 11 1544 110
XM_005250060.4 XP_005250117.1 10 - 11 1580 110
XM_011516629.2 XP_011514931.1 10 - 11 1536 110
XM_011516630.2 XP_011514932.1 10 - 11 1437 110
XM_011516631.2 XP_011514933.1 10 - 11 1565 110
XM_011516632.2 XP_011514934.1 10 - 11 1463 110
XM_017012723.1 XP_016868212.1 9 - 10 1300 110
GenBank mRNA AF062536.1 - 9 - 10 1318 110
AK096163.1 - 11 - 12 1432 110
AK096505.1 - 3 - 4 1203 110
AK296909.1 - 9 - 10 1269 110
AK315023.1 - 10 - 11 1720 110
BC125119.1 - 10 - 11 1457 110
BC125120.1 - 10 - 11 1457 110
BX537409.1 - 10 - 11 1474 110
U58087.1 - 10 - 11 1246 110

View Full Product Details