Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
-
Chromosome Location
Chr.7: 148698178 - 148801110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003592.2 NP_003583.2 13 - 14 1834 65
XM_005250060.4 XP_005250117.1 13 - 14 1870 65
XM_011516629.2 XP_011514931.1 13 - 14 1826 65
XM_011516630.2 XP_011514932.1 13 - 14 1727 65
XM_011516631.2 XP_011514933.1 13 - 14 1855 65
XM_011516632.2 XP_011514934.1 13 - 14 1753 65
XM_017012723.1 XP_016868212.1 12 - 13 1590 65
GenBank mRNA AF062536.1 - 12 - 13 1608 65
AK096163.1 - 14 - 15 1722 65
AK096505.1 - 6 - 7 1491 65
AK296909.1 - 12 - 13 1559 65
AK315023.1 - 13 - 14 2010 65
BC125119.1 - 13 - 14 1747 65
BC125120.1 - 13 - 14 1747 65
BX537409.1 - 13 - 14 1764 65
U58087.1 - 13 - 14 1536 65

View Full Product Details