The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003592.2 | NP_003583.2 | 14 - 15 | 1943 | 88 |
XM_005250060.4 | XP_005250117.1 | 14 - 15 | 1979 | 88 | |
XM_011516629.2 | XP_011514931.1 | 14 - 15 | 1935 | 88 | |
XM_011516630.2 | XP_011514932.1 | 14 - 15 | 1836 | 88 | |
XM_011516631.2 | XP_011514933.1 | 14 - 15 | 1964 | 88 | |
XM_011516632.2 | XP_011514934.1 | 14 - 15 | 1862 | 88 | |
XM_017012723.1 | XP_016868212.1 | 13 - 14 | 1699 | 88 | |
GenBank mRNA | AF062536.1 | - | 13 - 14 | 1717 | 88 |
AK096163.1 | - | 15 - 16 | 1831 | 88 | |
AK096505.1 | - | 7 - 8 | 1600 | 88 | |
AK296909.1 | - | 13 - 14 | 1668 | 88 | |
AK315023.1 | - | 14 - 15 | 2119 | 88 | |
BC125119.1 | - | 14 - 15 | 1856 | 88 | |
BC125120.1 | - | 14 - 15 | 1856 | 88 | |
BX537409.1 | - | 14 - 15 | 1873 | 88 | |
U58087.1 | - | 14 - 15 | 1645 | 88 |