Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
-
Chromosome Location
Chr.7: 148698178 - 148801110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011516631.2 XP_011514933.1 1 - 2 209 140

View Full Product Details