Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUBGCP5
Entrez Gene ID
114791
Gene Name
tubulin gamma complex associated protein 5
Gene Aliases
GCP5
Chromosome Location
Chr.15: 22982977 - 23039673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102610.1 NP_001096080.1 10 - 11 1290 109
NM_052903.4 NP_443135.3 10 - 11 1290 109
XM_005272540.2 XP_005272597.1 10 - 11 1197 109
XM_011543868.1 XP_011542170.1 10 - 11 1197 109
XM_011543869.2 XP_011542171.1 10 - 11 1194 109
XM_011543870.1 XP_011542172.1 10 - 11 1197 109
XM_017021890.1 XP_016877379.1 10 - 11 1197 109
XM_017021891.1 XP_016877380.1 10 - 11 1194 109
XM_017021892.1 XP_016877381.1 10 - 11 1194 109
XM_017021893.1 XP_016877382.1 10 - 11 1197 109
XM_017021894.1 XP_016877383.1 7 - 8 845 109
XM_017021895.1 XP_016877384.1 10 - 11 1214 109
XM_017021896.1 XP_016877385.1 2 - 3 329 109
XM_017021898.1 XP_016877387.1 5 - 6 604 109
XM_017021899.1 XP_016877388.1 10 - 11 1189 109
XM_017021900.1 XP_016877389.1 9 - 10 1064 109
GenBank mRNA AB067486.2 - 10 - 11 1204 109
AF272884.1 - 10 - 11 1214 109
AK056416.1 - 10 - 11 1189 109
AK289859.1 - 10 - 11 1189 109
AK310897.1 - 10 - 11 1186 109
BC071560.1 - 10 - 11 1186 109

View Full Product Details