The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102610.1 | NP_001096080.1 | 16 - 17 | 2444 | 107 |
NM_052903.4 | NP_443135.3 | 16 - 17 | 2444 | 107 | |
XM_005272540.2 | XP_005272597.1 | 16 - 17 | 2351 | 107 | |
XM_011543868.1 | XP_011542170.1 | 16 - 17 | 2351 | 107 | |
XM_011543869.2 | XP_011542171.1 | 16 - 17 | 2348 | 107 | |
XM_011543870.1 | XP_011542172.1 | 16 - 17 | 2351 | 107 | |
XM_017021890.1 | XP_016877379.1 | 16 - 17 | 2351 | 107 | |
XM_017021891.1 | XP_016877380.1 | 16 - 17 | 2348 | 107 | |
XM_017021892.1 | XP_016877381.1 | 16 - 17 | 2348 | 107 | |
XM_017021893.1 | XP_016877382.1 | 16 - 17 | 2351 | 107 | |
XM_017021894.1 | XP_016877383.1 | 13 - 14 | 1999 | 107 | |
XM_017021895.1 | XP_016877384.1 | 16 - 17 | 2368 | 107 | |
XM_017021896.1 | XP_016877385.1 | 8 - 9 | 1483 | 107 | |
XM_017021897.1 | XP_016877386.1 | 7 - 8 | 1243 | 107 | |
XM_017021898.1 | XP_016877387.1 | 11 - 12 | 1804 | 107 | |
XM_017021899.1 | XP_016877388.1 | 16 - 17 | 2389 | 107 | |
XM_017021900.1 | XP_016877389.1 | 15 - 16 | 2264 | 107 | |
XM_017021901.1 | XP_016877390.1 | 7 - 8 | 2313 | 107 | |
GenBank mRNA | AB067486.2 | - | 16 - 17 | 2358 | 107 |
AF272884.1 | - | 16 - 17 | 2368 | 107 | |
AK056416.1 | - | 16 - 17 | 2344 | 107 | |
AK289859.1 | - | 16 - 17 | 2343 | 107 | |
AK310897.1 | - | 16 - 17 | 2340 | 107 | |
BC046182.1 | - | 5 - 6 | 869 | 107 | |
BC071560.1 | - | 16 - 17 | 2340 | 107 |