Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TUBGCP5
- Entrez Gene ID
-
114791
- Gene Name
- tubulin gamma complex associated protein 5
- Gene Aliases
-
GCP5
- Chromosome Location
-
Chr.15: 22982977 - 23039673 on Build GRCh38
View Full Product Details