The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102610.1 | NP_001096080.1 | 9 - 10 | 1049 | 93 |
NM_052903.4 | NP_443135.3 | 9 - 10 | 1049 | 93 | |
XM_005272540.2 | XP_005272597.1 | 9 - 10 | 956 | 93 | |
XM_011543868.1 | XP_011542170.1 | 9 - 10 | 956 | 93 | |
XM_011543869.2 | XP_011542171.1 | 9 - 10 | 953 | 93 | |
XM_011543870.1 | XP_011542172.1 | 9 - 10 | 956 | 93 | |
XM_017021890.1 | XP_016877379.1 | 9 - 10 | 956 | 93 | |
XM_017021891.1 | XP_016877380.1 | 9 - 10 | 953 | 93 | |
XM_017021892.1 | XP_016877381.1 | 9 - 10 | 953 | 93 | |
XM_017021893.1 | XP_016877382.1 | 9 - 10 | 956 | 93 | |
XM_017021894.1 | XP_016877383.1 | 6 - 7 | 604 | 93 | |
XM_017021895.1 | XP_016877384.1 | 9 - 10 | 973 | 93 | |
XM_017021898.1 | XP_016877387.1 | 4 - 5 | 363 | 93 | |
XM_017021899.1 | XP_016877388.1 | 9 - 10 | 948 | 93 | |
GenBank mRNA | AB067486.2 | - | 9 - 10 | 963 | 93 |
AF272884.1 | - | 9 - 10 | 973 | 93 | |
AK056416.1 | - | 9 - 10 | 948 | 93 | |
AK289859.1 | - | 9 - 10 | 948 | 93 | |
AK310897.1 | - | 9 - 10 | 945 | 93 | |
BC071560.1 | - | 9 - 10 | 945 | 93 |