The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032998.1 | NP_001028170.1 | 10 - 11 | 1032 | 93 |
XM_017005217.1 | XP_016860706.1 | 10 - 11 | 994 | 93 | |
GenBank mRNA | BC000879.1 | - | 10 - 11 | 1001 | 93 |