Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY1
Entrez Gene ID
57590
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
FENS-1, WDF1, ZFYVE17
Chromosome Location
Chr.2: 223875343 - 223945387 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020830.4 NP_065881.1 1 - 2 243 68
GenBank mRNA AB037856.1 - 1 - 2 156 68
AF411977.1 - 1 - 2 140 68
AJ310568.1 - 1 - 2 230 68
AK022888.1 - 1 - 2 169 68
BC040525.1 - 1 - 2 155 68
BC065934.1 - 1 - 2 187 68
DA775670.1 - 1 - 2 243 68

View Full Product Details