The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032567.3 | NP_115956.3 | 1 - 1 | 144 | 84 |
GenBank mRNA | AF333098.1 | - | 1 - 1 | 115 | 84 |
AK098575.1 | - | 1 - 1 | 141 | 84 | |
AK314007.1 | - | 1 - 1 | 143 | 84 | |
BC033798.1 | - | 1 - 1 | 91 | 84 | |
BG723698.1 | - | 1 - 1 | 152 | 84 | |
BU853967.1 | - | 1 - 1 | 114 | 84 |