The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021629.3 | NP_067642.1 | 9 - 10 | 1198 | 96 |
XM_005247692.2 | XP_005247749.1 | 9 - 10 | 1119 | 96 | |
XM_006713721.2 | XP_006713784.1 | 9 - 10 | 1129 | 96 | |
GenBank mRNA | AF300648.1 | - | 8 - 9 | 918 | 96 |
AK001890.1 | - | 9 - 10 | 1198 | 96 | |
AK290571.1 | - | 9 - 10 | 1129 | 96 | |
AK297174.1 | - | 9 - 10 | 995 | 96 | |
BC000873.1 | - | 9 - 10 | 1008 | 96 |