The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006617.1 | NP_001006618.1 | 4 - 5 | 829 | 79 |
NM_001006618.1 | NP_001006619.1 | 4 - 5 | 829 | 79 | |
NM_001006619.1 | NP_001006620.1 | 4 - 5 | 829 | 79 | |
NM_001006620.1 | NP_001006621.1 | 3 - 4 | 501 | 79 | |
NM_001006621.1 | NP_001006622.1 | 2 - 3 | 411 | 79 | |
NM_024117.3 | NP_077022.1 | 4 - 5 | 829 | 79 | |
XM_017015126.1 | XP_016870615.1 | 4 - 5 | 718 | 79 | |
XM_017015127.1 | XP_016870616.1 | 4 - 5 | 718 | 79 | |
GenBank mRNA | AK290060.1 | - | 4 - 5 | 642 | 79 |
AK298841.1 | - | 1 - 2 | 80 | 79 | |
AL833042.1 | - | 3 - 4 | 462 | 79 | |
AY524429.1 | - | 4 - 5 | 826 | 79 | |
AY524430.1 | - | 4 - 5 | 826 | 79 | |
AY524431.1 | - | 4 - 5 | 826 | 79 | |
AY524432.1 | - | 4 - 5 | 826 | 79 | |
AY633624.1 | - | 4 - 5 | 796 | 79 | |
AY633625.1 | - | 4 - 5 | 803 | 79 | |
AY633626.1 | - | 4 - 5 | 803 | 79 | |
BC002326.2 | - | 4 - 5 | 798 | 79 | |
BC003044.1 | - | 4 - 5 | 798 | 79 | |
M37191.1 | - | 3 - 4 | 274 | 79 |