Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 28 - 29 4433 72
NM_001322988.1 NP_001309917.1 28 - 29 4433 72
NM_001322989.1 NP_001309918.1 28 - 29 4406 72
NM_001322990.1 NP_001309919.1 28 - 29 4394 72
NM_001322991.1 NP_001309920.1 28 - 29 4367 72
NM_001322992.1 NP_001309921.1 27 - 28 4283 72
NM_003947.5 NP_003938.1 28 - 29 4433 72
XM_006713810.3 XP_006713873.1 28 - 29 4678 72
XM_006713811.3 XP_006713874.1 28 - 29 4651 72
XM_006713812.3 XP_006713875.1 28 - 29 4639 72
XM_006713813.3 XP_006713876.1 28 - 29 4612 72
XM_006713814.3 XP_006713877.1 28 - 29 4678 72
XM_006713815.3 XP_006713878.1 28 - 29 4681 72
XM_011513279.2 XP_011511581.1 28 - 29 4678 72
XM_011513280.2 XP_011511582.1 28 - 29 4679 72
XM_011513281.2 XP_011511583.1 28 - 29 27300 72
XM_011513283.2 XP_011511585.1 28 - 29 4675 72
XM_011513285.2 XP_011511587.1 28 - 29 4674 72
XM_017007429.1 XP_016862918.1 28 - 29 4678 72
XM_017007430.1 XP_016862919.1 28 - 29 4678 72
XM_017007431.1 XP_016862920.1 28 - 29 4674 72
XM_017007432.1 XP_016862921.1 28 - 29 4674 72
XM_017007433.1 XP_016862922.1 28 - 29 4654 72
XM_017007434.1 XP_016862923.1 28 - 29 4642 72
XM_017007435.1 XP_016862924.1 28 - 29 4615 72
GenBank mRNA AK131379.1 - 18 - 19 2494 72
AK314126.1 - 28 - 29 4406 72
U94190.1 - 28 - 29 4376 72

View Full Product Details