Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 39 - 40 6009 115
NM_001322988.1 NP_001309917.1 39 - 40 6006 115
NM_001322993.1 NP_001309922.1 6 - 7 924 115
NM_001322994.1 NP_001309923.1 6 - 7 921 115
NM_001322995.1 NP_001309924.1 6 - 7 924 115
NM_001322996.1 NP_001309925.1 6 - 7 924 115
NM_001322997.1 NP_001309926.1 6 - 7 921 115
NM_001322998.1 NP_001309927.1 5 - 6 828 115
NM_001322999.1 NP_001309928.1 5 - 6 825 115
NM_001323000.1 NP_001309929.1 5 - 6 828 115
NM_001323001.1 NP_001309930.1 5 - 6 825 115
NM_007064.4 NP_008995.2 6 - 7 924 115
XM_006713810.3 XP_006713873.1 39 - 40 6254 115
XM_006713811.3 XP_006713874.1 39 - 40 6227 115
XM_006713812.3 XP_006713875.1 39 - 40 6215 115
XM_006713813.3 XP_006713876.1 39 - 40 6188 115
XM_006713814.3 XP_006713877.1 38 - 39 6158 115
XM_011513279.2 XP_011511581.1 39 - 40 6254 115
XM_011513280.2 XP_011511582.1 39 - 40 6252 115
XM_011513281.2 XP_011511583.1 39 - 40 28876 115
XM_011513283.2 XP_011511585.1 39 - 40 6251 115
XM_011513285.2 XP_011511587.1 38 - 39 6154 115
XM_017007429.1 XP_016862918.1 39 - 40 6254 115
XM_017007430.1 XP_016862919.1 38 - 39 6155 115
XM_017007431.1 XP_016862920.1 39 - 40 6247 115
XM_017007432.1 XP_016862921.1 38 - 39 6151 115
GenBank mRNA AB011422.1 - 6 - 7 878 115
AK125979.1 - 6 - 7 1039 115
AK296657.1 - 3 - 4 332 115

View Full Product Details