Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
Chromosome Location
Chr.1: 94820342 - 94895247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114106.2 NP_001107578.1 10 - 11 1343 86
NM_001258340.1 NP_001245269.1 10 - 11 1343 86
NM_001258341.1 NP_001245270.1 10 - 11 1247 86
NM_001258342.1 NP_001245271.1 9 - 10 1235 86
NM_001258343.1 NP_001245272.1 9 - 10 1139 86
NM_001301079.1 NP_001288008.1 9 - 10 1182 86
NM_152369.4 NP_689582.2 9 - 10 1278 86
XM_005270440.2 XP_005270497.1 10 - 11 1238 86
XM_005270441.2 XP_005270498.1 10 - 11 1280 86
XM_005270442.1 XP_005270499.1 10 - 11 1334 86
XM_011540632.2 XP_011538934.1 9 - 10 1207 86
XM_011540633.2 XP_011538935.1 9 - 10 1045 86
XM_011540635.2 XP_011538937.1 10 - 11 1334 86
XM_011540636.2 XP_011538938.1 10 - 11 1238 86
XM_017000262.1 XP_016855751.1 9 - 10 1220 86
XM_017000263.1 XP_016855752.1 10 - 11 1280 86
XM_017000264.1 XP_016855753.1 10 - 11 1334 86
XM_017000265.1 XP_016855754.1 10 - 11 1238 86
XM_017000266.1 XP_016855755.1 10 - 11 1334 86
GenBank mRNA AK300193.1 - 9 - 10 1076 86
AK301288.1 - 10 - 11 1084 86
AK302381.1 - 7 - 8 927 86
AK303906.1 - 9 - 10 1214 86
AK313574.1 - 10 - 11 1231 86
AY358659.1 - 6 - 7 797 86
BC033858.1 - 10 - 11 1306 86
BC053877.1 - 9 - 10 1201 86
BX648225.1 - 9 - 10 1207 86

View Full Product Details