Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
Chromosome Location
Chr.2: 189674292 - 189763059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144708.3 NP_653309.3 18 - 19 3665 172
XM_011510673.2 XP_011508975.1 18 - 19 4184 172
XM_011510674.1 XP_011508976.1 18 - 19 4151 172
XM_011510675.2 XP_011508977.1 17 - 18 4079 172
XM_011510676.2 XP_011508978.1 18 - 19 4184 172
XM_011510677.2 XP_011508979.1 18 - 19 4184 172
XM_011510679.2 XP_011508981.1 16 - 17 3755 172
XM_011510680.1 XP_011508982.1 18 - 19 4184 172
XM_011510681.1 XP_011508983.1 18 - 19 4184 172
XM_011510682.2 XP_011508984.1 18 - 19 4184 172
XM_011510685.2 XP_011508987.1 18 - 19 3740 172
XM_011510687.2 XP_011508989.1 17 - 18 3548 172
XM_011510691.2 XP_011508993.1 14 - 15 2370 172
XM_011510692.2 XP_011508994.1 16 - 17 3384 172
XM_011510693.2 XP_011508995.1 15 - 16 3280 172
XM_011510694.2 XP_011508996.1 14 - 15 3099 172
XM_017003413.1 XP_016858902.1 18 - 19 4184 172
XM_017003415.1 XP_016858904.1 13 - 14 2879 172
XM_017003416.1 XP_016858905.1 15 - 16 3291 172
XM_017003418.1 XP_016858907.1 13 - 14 2994 172
XM_017003419.1 XP_016858908.1 13 - 14 2961 172
XM_017003420.1 XP_016858909.1 13 - 14 2897 172
GenBank mRNA AJ549812.1 - 17 - 18 3364 172
AJ605761.1 - 6 - 7 833 172
AK058144.1 - 6 - 7 1017 172
AK126073.1 - 16 - 17 3435 172
BC037215.1 - 12 - 13 2654 172
BC044907.1 - 14 - 15 3158 172
BC047413.1 - 21 - 22 3229 172

View Full Product Details