Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
Chromosome Location
Chr.2: 189674292 - 189763059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011510677.2 XP_011508979.1 21 - 22 4619 96
GenBank mRNA BC044907.1 - 17 - 18 3593 96

View Full Product Details