Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA10
Entrez Gene ID
57053
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Gene Aliases
-
Chromosome Location
Chr.11: 3665587 - 3673208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303034.1 NP_001289963.1 4 - 5 1028 135
NM_001303035.1 NP_001289964.1 4 - 5 961 135
NM_020402.3 NP_065135.2 4 - 5 966 135
XM_011520234.1 XP_011518536.1 3 - 4 4162 135
GenBank mRNA AF199235.2 - 4 - 5 966 135
AJ278118.1 - 4 - 5 135
AJ295237.1 - 4 - 5 936 135
BC136250.1 - 4 - 5 944 135
BC143303.1 - 4 - 5 1011 135

View Full Product Details