The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004444.4 | NP_004435.3 | 16 - 17 | 3322 | 73 |
XM_017011816.1 | XP_016867305.1 | 16 - 17 | 2979 | 73 | |
GenBank mRNA | AB209644.1 | - | 15 - 16 | 4032 | 73 |
AK301224.1 | - | 11 - 12 | 1629 | 73 | |
BC004264.1 | - | 13 - 14 | 2407 | 73 | |
BC052804.1 | - | 16 - 17 | 2999 | 73 | |
U07695.1 | - | 16 - 17 | 2916 | 73 |