The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004444.4 | NP_004435.3 | 2 - 3 | 609 | 64 |
XM_017011816.1 | XP_016867305.1 | 2 - 3 | 212 | 64 | |
GenBank mRNA | AB209644.1 | - | 1 - 2 | 1319 | 64 |
AY056048.1 | - | 2 - 3 | 203 | 64 | |
BC052804.1 | - | 2 - 3 | 286 | 64 | |
EU826608.1 | - | 2 - 3 | 118 | 64 | |
EU826609.1 | - | 2 - 3 | 118 | 64 | |
EU826610.1 | - | 2 - 3 | 118 | 64 | |
U07695.1 | - | 2 - 3 | 203 | 64 |