The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004444.4 | NP_004435.3 | 9 - 10 | 2179 | 64 |
XM_017011816.1 | XP_016867305.1 | 9 - 10 | 1836 | 64 | |
GenBank mRNA | AB209644.1 | - | 8 - 9 | 2889 | 64 |
AK301224.1 | - | 4 - 5 | 486 | 64 | |
AY056048.1 | - | 9 - 10 | 1773 | 64 | |
BC004264.1 | - | 6 - 7 | 1264 | 64 | |
BC052804.1 | - | 9 - 10 | 1856 | 64 | |
U07695.1 | - | 9 - 10 | 1773 | 64 |