The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018073.7 | NP_060543.5 | 1 - 2 | 230 | 90 |
GenBank mRNA | AF360739.1 | - | 1 - 2 | 183 | 90 |
AK056523.1 | - | 1 - 2 | 192 | 90 | |
AK291166.1 | - | 1 - 2 | 142 | 90 | |
AL832730.1 | - | 1 - 2 | 180 | 90 | |
BC109063.2 | - | 1 - 2 | 149 | 90 | |
DC402744.1 | - | 1 - 2 | 230 | 90 |