Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 4 - 5 578 70
NM_001141974.2 NP_001135446.1 4 - 5 578 70
NM_022089.3 NP_071372.1 4 - 5 578 70
XM_005245810.1 XP_005245867.1 4 - 5 578 70
XM_005245811.1 XP_005245868.1 4 - 5 578 70
XM_005245812.1 XP_005245869.1 4 - 5 578 70
XM_005245815.1 XP_005245872.1 4 - 5 578 70
XM_006710512.1 XP_006710575.1 4 - 5 578 70
XM_006710513.1 XP_006710576.1 4 - 5 578 70
XM_011541128.1 XP_011539430.1 4 - 5 578 70
XM_011541129.1 XP_011539431.1 4 - 5 578 70
XM_017000844.1 XP_016856333.1 4 - 5 578 70
XM_017000845.1 XP_016856334.1 4 - 5 578 70
XM_017000846.1 XP_016856335.1 4 - 5 578 70
XM_017000847.1 XP_016856336.1 4 - 5 578 70
XM_017000848.1 XP_016856337.1 4 - 5 578 70
XM_017000849.1 XP_016856338.1 4 - 5 578 70
XM_017000850.1 XP_016856339.1 4 - 5 578 70
GenBank mRNA AK290210.1 - 4 - 5 534 70
AL354615.1 - 4 - 5 378 70
AY461712.1 - 4 - 5 392 70
BC030267.1 - 4 - 5 521 70
BC034575.1 - 1 - 2 69 70

View Full Product Details