The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136204.2 | NP_001129676.1 | 11 - 12 | 1517 | 93 |
NM_018715.3 | NP_061185.1 | 12 - 13 | 1686 | 93 | |
GenBank mRNA | AB040903.1 | - | 12 - 13 | 1617 | 93 |
AJ421269.1 | - | 12 - 13 | 1703 | 93 | |
AL359612.1 | - | 9 - 10 | 1035 | 93 | |
BC004933.1 | - | 8 - 9 | 957 | 93 | |
BC042141.1 | - | 11 - 12 | 1482 | 93 | |
BC053908.1 | - | 11 - 12 | 1515 | 93 | |
BC142946.1 | - | 11 - 12 | 1273 | 93 |