The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313726.1 | NP_001300655.1 | 24 - 25 | 2666 | 63 |
NM_001313727.1 | NP_001300656.1 | 20 - 21 | 2224 | 63 | |
NM_031418.3 | NP_113606.2 | 23 - 24 | 2574 | 63 | |
XM_011520282.2 | XP_011518584.1 | 10 - 11 | 1121 | 63 | |
XM_017018118.1 | XP_016873607.1 | 21 - 22 | 2202 | 63 | |
XM_017018119.1 | XP_016873608.1 | 18 - 19 | 1814 | 63 | |
GenBank mRNA | AJ300461.1 | - | 23 - 24 | 3281 | 63 |
AK295658.1 | - | 7 - 8 | 609 | 63 | |
AK295816.1 | - | 20 - 21 | 2136 | 63 | |
AK302659.1 | - | 23 - 24 | 2613 | 63 | |
AK304781.1 | - | 24 - 25 | 2507 | 63 | |
AK316018.1 | - | 20 - 21 | 2136 | 63 | |
BQ430769.1 | - | 4 - 5 | 445 | 63 |