Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 26 - 27 2894 60
NM_001313727.1 NP_001300656.1 22 - 23 2452 60
NM_031418.3 NP_113606.2 25 - 26 2802 60
XM_011520282.2 XP_011518584.1 12 - 13 1349 60
XM_017018118.1 XP_016873607.1 23 - 24 2430 60
XM_017018119.1 XP_016873608.1 20 - 21 2042 60
GenBank mRNA AJ300461.1 - 25 - 26 3509 60
AK295658.1 - 9 - 10 837 60
AK295816.1 - 22 - 23 2364 60
AK302659.1 - 25 - 26 2841 60
AK304781.1 - 26 - 27 2735 60
AK316018.1 - 22 - 23 2364 60

View Full Product Details