Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH15
Entrez Gene ID
65217
Gene Name
protocadherin related 15
Gene Aliases
CDHR15, DFNB23, USH1F
Chromosome Location
Chr.10: 53802771 - 55629182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142763.1 NP_001136235.1 13 - 14 1850 69
NM_001142764.1 NP_001136236.1 12 - 13 1835 69
NM_001142765.1 NP_001136237.1 12 - 13 1835 69
NM_001142766.1 NP_001136238.1 12 - 13 1835 69
NM_001142767.1 NP_001136239.1 11 - 12 1724 69
NM_001142768.1 NP_001136240.1 11 - 12 1769 69
NM_001142769.1 NP_001136241.1 14 - 15 1871 69
NM_001142770.1 NP_001136242.1 12 - 13 1835 69
NM_001142771.1 NP_001136243.1 13 - 14 1850 69
NM_001142772.1 NP_001136244.1 12 - 13 1835 69
NM_001142773.1 NP_001136245.1 11 - 12 1769 69
NM_033056.3 NP_149045.3 12 - 13 1835 69
XM_017016571.1 XP_016872060.1 12 - 13 2433 69
XM_017016572.1 XP_016872061.1 12 - 13 2431 69
XM_017016573.1 XP_016872062.1 19 - 20 5232 69
GenBank mRNA AK126923.1 - 14 - 15 1736 69
AL834134.1 - 12 - 13 1732 69
AY029205.1 - 12 - 13 1823 69
AY029237.1 - 12 - 13 1835 69
EU718480.1 - 13 - 14 1856 69
EU718481.1 - 12 - 13 1835 69
EU718482.1 - 12 - 13 1835 69

View Full Product Details