The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142763.1 | NP_001136235.1 | 29 - 30 | 4212 | 89 |
NM_001142764.1 | NP_001136236.1 | 28 - 29 | 4197 | 89 | |
NM_001142765.1 | NP_001136237.1 | 26 - 27 | 3984 | 89 | |
NM_001142766.1 | NP_001136238.1 | 28 - 29 | 4197 | 89 | |
NM_001142767.1 | NP_001136239.1 | 27 - 28 | 4086 | 89 | |
NM_001142768.1 | NP_001136240.1 | 27 - 28 | 4131 | 89 | |
NM_001142769.1 | NP_001136241.1 | 30 - 31 | 4233 | 89 | |
NM_001142770.1 | NP_001136242.1 | 28 - 29 | 4197 | 89 | |
NM_001142771.1 | NP_001136243.1 | 29 - 30 | 4212 | 89 | |
NM_001142772.1 | NP_001136244.1 | 28 - 29 | 4197 | 89 | |
NM_001142773.1 | NP_001136245.1 | 27 - 28 | 4131 | 89 | |
NM_033056.3 | NP_149045.3 | 28 - 29 | 4197 | 89 | |
XM_017016571.1 | XP_016872060.1 | 28 - 29 | 4795 | 89 | |
XM_017016572.1 | XP_016872061.1 | 28 - 29 | 4793 | 89 | |
XM_017016573.1 | XP_016872062.1 | 35 - 36 | 7594 | 89 | |
GenBank mRNA | AY029205.1 | - | 28 - 29 | 4185 | 89 |
AY029237.1 | - | 28 - 29 | 4197 | 89 | |
AY388963.1 | - | 5 - 6 | 821 | 89 | |
EU718480.1 | - | 29 - 30 | 4218 | 89 | |
EU718481.1 | - | 28 - 29 | 4197 | 89 | |
EU718482.1 | - | 28 - 29 | 4197 | 89 |