Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TREM1
Entrez Gene ID
54210
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
CD354, TREM-1
Chromosome Location
Chr.6: 41267385 - 41286745 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514696.1 XP_011512998.1 3 - 4 633 80
GenBank mRNA BI910026.1 - 3 - 4 613 80

View Full Product Details