The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007270.4 | NP_009201.2 | 1 - 2 | 385 | 101 |
XM_011515115.2 | XP_011513417.1 | 1 - 2 | 385 | 101 | |
GenBank mRNA | AF089745.1 | - | 1 - 2 | 56 | 101 |
AK075331.1 | - | 1 - 2 | 349 | 101 | |
AK128597.1 | - | 2 - 3 | 326 | 101 | |
AK293954.1 | - | 1 - 2 | 315 | 101 | |
BC101723.1 | - | 1 - 2 | 313 | 101 | |
BC112053.1 | - | 1 - 2 | 313 | 101 | |
DA234297.1 | - | 1 - 2 | 385 | 101 |