The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319659.1 | NP_001306588.1 | 1 - 2 | 62 | 59 |
GenBank mRNA | AK000449.1 | - | 1 - 2 | 39 | 59 |
BQ679163.1 | - | 1 - 2 | 74 | 59 |