Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGLYRP4
Entrez Gene ID
57115
Gene Name
peptidoglycan recognition protein 4
Gene Aliases
PGLYRPIbeta, PGRP-Ibeta, PGRPIB, SBBI67
Chromosome Location
Chr.1: 153327410 - 153348844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020393.3 NP_065126.2 1 - 2 319 117
XM_011509790.1 XP_011508092.1 1 - 2 319 117
XM_011509791.2 XP_011508093.1 1 - 2 322 117
XM_011509792.1 XP_011508094.1 1 - 2 319 117
GenBank mRNA AK292203.1 - 1 - 2 117
BC142636.1 - 1 - 2 294 117

View Full Product Details