Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGLYRP4
Entrez Gene ID
57115
Gene Name
peptidoglycan recognition protein 4
Gene Aliases
PGLYRPIbeta, PGRP-Ibeta, PGRPIB, SBBI67
Chromosome Location
Chr.1: 153327410 - 153348844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011509791.2 XP_011508093.1 3 - 4 508 82
GenBank mRNA AF242518.1 - 3 - 4 233 82

View Full Product Details