The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020393.3 | NP_065126.2 | 7 - 8 | 1184 | 65 |
XM_011509789.2 | XP_011508091.1 | 7 - 8 | 1196 | 65 | |
XM_011509790.1 | XP_011508092.1 | 7 - 8 | 1184 | 65 | |
XM_011509791.2 | XP_011508093.1 | 7 - 8 | 1175 | 65 | |
XM_011509792.1 | XP_011508094.1 | 7 - 8 | 1184 | 65 | |
XM_011509793.2 | XP_011508095.1 | 4 - 5 | 872 | 65 | |
GenBank mRNA | AF242518.1 | - | 7 - 8 | 897 | 65 |
AK292203.1 | - | 7 - 8 | 1187 | 65 | |
AY035377.1 | - | 7 - 8 | 889 | 65 | |
BC107157.1 | - | 6 - 7 | 871 | 65 | |
BC107158.1 | - | 6 - 7 | 871 | 65 | |
BC142636.1 | - | 7 - 8 | 1159 | 65 |