The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153366.3 | NP_699197.3 | 12 - 13 | 2697 | 105 |
GenBank mRNA | AJ619977.1 | - | 12 - 13 | 2523 | 105 |
AK122605.1 | - | 12 - 13 | 2693 | 105 | |
AY243503.1 | - | 12 - 13 | 2523 | 105 | |
BC030816.1 | - | 12 - 13 | 2697 | 105 |