The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144030.1 | NP_001137502.1 | 16 - 17 | 1937 | 58 |
NM_024662.2 | NP_078938.2 | 18 - 19 | 2152 | 58 | |
GenBank mRNA | AB051496.1 | - | 18 - 19 | 2073 | 58 |
AF489535.1 | - | 14 - 15 | 1449 | 58 | |
AK001636.1 | - | 18 - 19 | 2007 | 58 | |
AK022241.1 | - | 18 - 19 | 2114 | 58 | |
AK074430.1 | - | - | 180 | 58 | |
AK225341.1 | - | 18 - 19 | 2114 | 58 | |
AK294044.1 | - | 16 - 17 | 1836 | 58 | |
AL136882.1 | - | 18 - 19 | 2009 | 58 | |
BC035558.1 | - | 18 - 19 | 2086 | 58 |