The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204824.1 | NP_001191753.1 | 7 - 8 | 820 | 104 |
NM_004519.3 | NP_004510.1 | 7 - 8 | 1369 | 104 | |
XM_005250914.3 | XP_005250971.1 | 4 - 5 | 533 | 104 | |
XM_006716555.3 | XP_006716618.1 | 5 - 6 | 630 | 104 | |
XM_011517026.2 | XP_011515328.1 | 7 - 8 | 3903 | 104 | |
XM_017013400.1 | XP_016868889.1 | 7 - 8 | 10875 | 104 | |
GenBank mRNA | AB208890.1 | - | 7 - 8 | 922 | 104 |
AF033347.1 | - | 7 - 8 | 1003 | 104 | |
AK296293.1 | - | 7 - 8 | 820 | 104 | |
BC128576.1 | - | 8 - 9 | 1112 | 104 |