Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF3
Entrez Gene ID
6428
Gene Name
serine and arginine rich splicing factor 3
Gene Aliases
SFRS3, SRp20
Chromosome Location
Chr.6: 36594313 - 36604467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003017.4 NP_003008.1 2 - 3 380 69
NR_036610.1 2 - 3 380 69
GenBank mRNA AB451229.1 - 1 - 2 209 69
AB451352.1 - 1 - 2 209 69
AF107405.1 - 2 - 3 304 69
AK304097.1 - 2 - 3 327 69
AK310317.1 - - 958 69
AK312553.1 - 2 - 3 331 69
BC000914.1 - 2 - 3 314 69
BC069018.1 - 2 - 3 310 69
BT007017.1 - 1 - 2 209 69
DC398250.1 - 2 - 3 472 69
L10838.1 - 1 - 2 209 69

View Full Product Details