The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080751.2 | NP_542789.2 | 4 - 5 | 566 | 67 |
XM_005260660.3 | XP_005260717.1 | 2 - 3 | 760 | 67 | |
GenBank mRNA | AF417580.2 | - | 4 - 5 | 566 | 67 |
AK294013.1 | - | 2 - 3 | 312 | 67 |