Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMC2
Entrez Gene ID
117532
Gene Name
transmembrane channel like 2
Gene Aliases
C20orf145
Chromosome Location
Chr.20: 2536607 - 2642185 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080751.2 NP_542789.2 9 - 10 1089 63
XM_005260660.3 XP_005260717.1 7 - 8 1283 63
GenBank mRNA AF417580.2 - 9 - 10 1089 63
AK294013.1 - 7 - 8 835 63
AK316261.1 - 7 - 8 805 63

View Full Product Details