The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005215.3 | NP_005206.2 | 28 - 29 | 4873 | 69 |
XM_011525844.2 | XP_011524146.1 | 25 - 26 | 3761 | 69 | |
XM_017025568.1 | XP_016881057.1 | 28 - 29 | 4867 | 69 | |
XM_017025569.1 | XP_016881058.1 | 28 - 29 | 4813 | 69 | |
XM_017025570.1 | XP_016881059.1 | 24 - 25 | 3769 | 69 | |
GenBank mRNA | X76132.1 | - | 28 - 29 | 4257 | 69 |