The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005215.3 | NP_005206.2 | 4 - 5 | 1457 | 70 |
XM_011525843.1 | XP_011524145.1 | 4 - 5 | 1457 | 70 | |
XM_017025568.1 | XP_016881057.1 | 4 - 5 | 1457 | 70 | |
XM_017025569.1 | XP_016881058.1 | 4 - 5 | 1457 | 70 | |
GenBank mRNA | AK302643.1 | - | 4 - 5 | 762 | 70 |
BC036524.1 | - | 3 - 4 | 643 | 70 | |
M32292.1 | - | 4 - 5 | 847 | 70 | |
X76132.1 | - | 4 - 5 | 841 | 70 |