The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_136628.1 | 9 - 10 | 876 | 63 | |
NR_136630.1 | 9 - 10 | 1063 | 63 | ||
NR_136631.1 | 10 - 11 | 1143 | 63 | ||
GenBank mRNA | AK304488.1 | - | 10 - 11 | 1166 | 63 |