Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TASP1
Entrez Gene ID
55617
Gene Name
taspase 1
Gene Aliases
C20orf13, dJ585I14.2
Chromosome Location
Chr.20: 13104760 - 13638936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323602.1 NP_001310531.1 9 - 10 893 64
NM_001323603.1 NP_001310532.1 14 - 15 1553 64
NM_001323604.1 NP_001310533.1 14 - 15 1361 64
NM_017714.2 NP_060184.2 13 - 14 1286 64
NR_136628.1 12 - 13 1166 64
NR_136629.1 11 - 12 1099 64
NR_136630.1 12 - 13 1353 64
NR_136631.1 13 - 14 1433 64
XM_011529292.1 XP_011527594.1 15 - 16 1609 64
XM_011529294.1 XP_011527596.1 15 - 16 1623 64
XM_017027927.1 XP_016883416.1 17 - 18 1720 64
XM_017027928.1 XP_016883417.1 14 - 15 1212 64
XM_017027930.1 XP_016883419.1 12 - 13 1195 64
GenBank mRNA AK000219.1 - 12 - 13 1253 64
AK025671.1 - 12 - 13 1253 64
AK299940.1 - 7 - 8 686 64
BC025266.1 - 13 - 14 1266 64

View Full Product Details