Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TASP1
Entrez Gene ID
55617
Gene Name
taspase 1
Gene Aliases
C20orf13, dJ585I14.2
Chromosome Location
Chr.20: 13104760 - 13638936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323602.1 NP_001310531.1 3 - 4 326 95
NM_001323603.1 NP_001310532.1 3 - 4 326 95
NM_001323604.1 NP_001310533.1 3 - 4 326 95
NM_017714.2 NP_060184.2 3 - 4 326 95
NR_136628.1 3 - 4 326 95
NR_136629.1 3 - 4 326 95
NR_136630.1 3 - 4 326 95
NR_136631.1 3 - 4 326 95
XM_011529292.1 XP_011527594.1 3 - 4 307 95
XM_011529294.1 XP_011527596.1 3 - 4 307 95
XM_017027927.1 XP_016883416.1 3 - 4 307 95
XM_017027929.1 XP_016883418.1 3 - 4 322 95
XM_017027930.1 XP_016883419.1 3 - 4 320 95
XM_017027931.1 XP_016883420.1 3 - 4 322 95
GenBank mRNA AK000219.1 - 2 - 3 293 95
AK025671.1 - 2 - 3 293 95
AK299940.1 - 3 - 4 318 95
AK304488.1 - 3 - 4 322 95
AK316311.1 - 3 - 4 307 95
AL601028.1 - 3 - 4 330 95
BC025266.1 - 3 - 4 306 95

View Full Product Details