The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042417.1 | NP_001035882.1 | 9 - 10 | 940 | 93 |
NM_001278225.1 | NP_001265154.1 | 9 - 10 | 868 | 93 | |
NM_017987.4 | NP_060457.4 | 10 - 11 | 1369 | 93 | |
NR_103475.1 | 10 - 11 | 1217 | 93 | ||
NR_103476.1 | 10 - 11 | 1042 | 93 | ||
XM_005269953.4 | XP_005270010.1 | 9 - 10 | 1267 | 93 | |
XM_005269955.4 | XP_005270012.1 | 9 - 10 | 948 | 93 | |
XM_005269957.4 | XP_005270014.1 | 9 - 10 | 937 | 93 | |
XM_006717911.3 | XP_006717974.1 | 10 - 11 | 1040 | 93 | |
XM_011539942.2 | XP_011538244.1 | 10 - 11 | 1781 | 93 | |
GenBank mRNA | AB040970.1 | - | 10 - 11 | 978 | 93 |
AF411980.1 | - | 9 - 10 | 889 | 93 | |
AF461266.1 | - | 9 - 10 | 948 | 93 | |
AK127019.1 | - | 10 - 11 | 1369 | 93 | |
AK294211.1 | - | 9 - 10 | 851 | 93 | |
AY528716.1 | - | 10 - 11 | 1007 | 93 | |
BC041092.1 | - | 9 - 10 | 911 | 93 |