The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015043.3 | NP_055858.2 | 15 - 16 | 2526 | 81 |
NM_198868.2 | NP_942568.2 | 15 - 16 | 2526 | 81 | |
GenBank mRNA | AB014576.1 | - | 15 - 16 | 2526 | 81 |
AB449893.1 | - | 15 - 16 | 2489 | 81 | |
AK001196.1 | - | 2 - 3 | 139 | 81 | |
AK074791.1 | - | 5 - 6 | 608 | 81 | |
AK223509.1 | - | 1 - 2 | 77 | 81 | |
AK304757.1 | - | 1 - 2 | 360 | 81 | |
BC000701.2 | - | 2 - 3 | 160 | 81 | |
BC008919.2 | - | 4 - 5 | 508 | 81 | |
BC080659.1 | - | 4 - 5 | 587 | 81 | |
BC129994.1 | - | 15 - 16 | 2530 | 81 |