Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D9B
Entrez Gene ID
23061
Gene Name
TBC1 domain family member 9B
Gene Aliases
GRAMD9B
Chromosome Location
Chr.5: 179862071 - 179907856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015043.3 NP_055858.2 1 - 2 159 135
NM_198868.2 NP_942568.2 1 - 2 159 135
GenBank mRNA AB014576.1 - 1 - 2 159 135
AB449893.1 - 1 - 2 122 135
BC129994.1 - 1 - 2 163 135
BM459848.1 - - 174 135
BU183640.1 - 1 - 2 172 135

View Full Product Details