The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015043.3 | NP_055858.2 | 4 - 5 | 618 | 82 |
NM_198868.2 | NP_942568.2 | 4 - 5 | 618 | 82 | |
GenBank mRNA | AB014576.1 | - | 4 - 5 | 618 | 82 |
AB449893.1 | - | 4 - 5 | 581 | 82 | |
BC129994.1 | - | 4 - 5 | 622 | 82 | |
BU183640.1 | - | 4 - 5 | 632 | 82 |